Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRD5A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024592.4 | NP_078868.1 | 5 | 1034 | ||
XM_005265766.3 | XP_005265823.1 | 4 | 1006 | |||
XM_005265767.3 | XP_005265824.1 | 3 | 694 | |||
XM_017008601.1 | XP_016864090.1 | 5 | 809 | |||
GenBank mRNA | AK023414.1 | - | 5 | 1014 | ||
BC002480.2 | - | 5 | 1010 | 3350037 | ||
CR457312.1 | - | 5 | 918 |