Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278.4 | NP_001269.3 | 12 | 1342 | ||
NM_001320928.1 | NP_001307857.1 | 12 | 1342 | |||
XM_017015611.1 | XP_016871100.1 | 12 | 1342 | |||
XM_017015612.1 | XP_016871101.1 | 12 | 1342 | |||
XM_017015613.1 | XP_016871102.1 | 2 | 157 | |||
GenBank mRNA | AB209028.1 | - | 12 | 1296 | ||
AF009225.1 | - | 12 | 1299 | |||
AF012890.1 | - | 12 | 1264 | |||
AF080157.1 | - | 12 | 1359 | |||
BC092514.1 | - | 12 | 1280 | 3063613 | ||
BM800404.1 | - | 527 | ||||
U22512.1 | - | 11 | 1172 |