Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278.4 | NP_001269.3 | 12, 13 | 1432 | ||
NM_001320928.1 | NP_001307857.1 | 12, 13 | 1432 | |||
XM_017015611.1 | XP_016871100.1 | 12, 13 | 1432 | |||
XM_017015612.1 | XP_016871101.1 | 12, 13 | 1432 | |||
XM_017015613.1 | XP_016871102.1 | 2, 3 | 247 | |||
GenBank mRNA | AB209028.1 | - | 12, 13 | 1386 | ||
AF009225.1 | - | 12, 13 | 1389 | |||
AF012890.1 | - | 12, 13 | 1354 | |||
AF080157.1 | - | 12, 13 | 1449 | |||
BC092514.1 | - | 12, 13 | 1370 | 3063613 | ||
BM800404.1 | - | 617 | ||||
U22512.1 | - | 11, 12 | 1262 |