Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFTUD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142605.1 | NP_001136077.1 | 9 | 967 | ||
NM_001258353.1 | NP_001245282.1 | 10 | 1213 | |||
NM_001258354.1 | NP_001245283.1 | 10 | 1046 | |||
NM_004247.3 | NP_004238.3 | 10 | 1076 | |||
GenBank mRNA | AJ505017.1 | - | 6 | 450 | ||
AK126464.1 | - | 11 | 882 | |||
AK128148.1 | - | 10 | 1213 | |||
AK225335.1 | - | 10 | 836 | |||
AK293108.1 | - | 10 | 839 | |||
AK296367.1 | - | 9 | 770 | |||
AK297392.1 | - | 10 | 852 | |||
AK302828.1 | - | 10 | 716 | |||
AK316098.1 | - | 9 | 787 | |||
BC002360.2 | - | 10 | 846 | 2960986 | ||
CR456774.1 | - | 9 | 815 | |||
D21163.1 | - | 10 | 875 |