Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFTUD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142605.1 | NP_001136077.1 | 17 | 1911 | ||
NM_001258353.1 | NP_001245282.1 | 18 | 2157 | |||
NM_001258354.1 | NP_001245283.1 | 18 | 1990 | |||
NM_004247.3 | NP_004238.3 | 18 | 2020 | |||
GenBank mRNA | AJ505017.1 | - | 14 | 1394 | ||
AK126193.1 | - | 8 | 1487 | |||
AK126464.1 | - | 19 | 1826 | |||
AK126652.1 | - | 2 | 1418 | |||
AK128148.1 | - | 18 | 2157 | |||
AK225335.1 | - | 18 | 1780 | |||
AK293108.1 | - | 18 | 1783 | |||
AK296367.1 | - | 17 | 1714 | |||
AK297392.1 | - | 18 | 1796 | |||
AK302828.1 | - | 18 | 1660 | |||
AK316098.1 | - | 17 | 1731 | |||
BC002360.2 | - | 18 | 1790 | 2960986 | ||
CR456774.1 | - | 17 | 1759 | |||
D21163.1 | - | 18 | 1819 |