Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006446.4 | NP_006437.3 | 5 | 523 | ||
GenBank mRNA | AB026257.1 | - | 5 | 518 | ||
AF060500.1 | - | 4 | 419 | |||
AF205071.1 | - | 5 | 553 | |||
AJ132573.1 | - | 4 | 482 | |||
AK312970.1 | - | 5 | 505 | |||
BC005243.2 | - | 4 | 481 | |||
BC020696.1 | - | 5 | 524 | |||
BC054878.1 | - | 5 | 511 | |||
BC070172.1 | - | 5 | 521 | |||
BC105596.1 | - | 5 | 511 |