Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164675.1 | NP_001158147.1 | 3 | 546 | ||
NM_182760.3 | NP_877437.2 | 3 | 546 | |||
XM_011533624.2 | XP_011531926.1 | 3 | 537 | |||
XM_011533625.2 | XP_011531927.1 | 3 | 537 | |||
XM_011533626.2 | XP_011531928.1 | 3 | 537 | |||
XM_017006252.1 | XP_016861741.1 | 3 | 537 | |||
XM_017006254.1 | XP_016861743.1 | 3 | 537 | |||
XM_017006255.1 | XP_016861744.1 | 3 | 537 | |||
GenBank mRNA | AB448737.1 | - | 3 | 517 | ||
AK057983.1 | - | 4 | 445 | |||
AK075459.1 | - | 3 | 528 | |||
AY208752.1 | - | 3 | 509 | |||
AY323910.1 | - | 3 | 509 | |||
AY358092.1 | - | 3 | 526 | |||
BC110862.1 | - | 3 | 517 | 5760490 | ||
BC121122.2 | - | 3 | 538 | 40120670 | ||
BC121123.2 | - | 3 | 538 | 40120671 | ||
BG705404.1 | - | 3 | 539 |