Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085479.2 | NP_001078948.1 | 7 | 865 | ||
NM_001207023.1 | NP_001193952.1 | 7 | 832 | |||
GenBank mRNA | BC146874.1 | - | 7 | 832 | 9056802 | |
BC146889.1 | - | 7 | 865 | 9056817 | ||
BC171811.1 | - | 7 | 832 | 9054465 | ||
BC171818.1 | - | 7 | 865 | 9054472 |