Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000118.3 | NP_000109.1 | 7 | 1259 | ||
NM_001114753.2 | NP_001108225.1 | 7 | 1259 | |||
NM_001278138.1 | NP_001265067.1 | 7 | 999 | |||
GenBank mRNA | AK222669.1 | - | 7 | 1172 | ||
AK290389.1 | - | 7 | 1143 | |||
AK301171.1 | - | 7 | 999 | |||
AK302455.1 | - | 4 | 371 | |||
BC014271.2 | - | 7 | 993 | 3843174 | ||
BT006872.1 | - | 7 | 841 | |||
J05481.1 | - | 7 | 802 | |||
X72012.1 | - | 7 | 1122 |