Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270523.1 | NP_001257452.1 | 5 | 1886 | ||
NM_001270524.1 | NP_001257453.1 | 4 | 2039 | |||
NM_001270525.1 | NP_001257454.1 | 3 | 1833 | |||
NM_021728.3 | NP_068374.1 | 5 | 1910 | |||
NM_172337.2 | NP_758840.1 | 3 | 1809 | |||
NR_073034.1 | 4 | 1734 | ||||
NR_073036.1 | 2 | 1657 | ||||
GenBank mRNA | AB593056.1 | - | 2 | 1660 | ||
AB593057.1 | - | 3 | 1579 | |||
AB593058.1 | - | 5 | 1910 | |||
BC032579.1 | - | 5 | 1911 | 5493541 |