Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000314.6 | NP_000305.3 | 8 | 1931 | ||
NM_001304717.2 | NP_001291646.2 | 8 | 1931 | |||
NM_001304718.1 | NP_001291647.1 | 8 | 2045 | |||
GenBank mRNA | AK313581.1 | - | 8 | 1092 | ||
BC005821.2 | - | 8 | 1736 | 3937787 | ||
CR450306.1 | - | 8 | 900 | |||
JF268690.1 | - | 8 | 1063 | |||
KJ534922.1 | - | 8 | 900 | |||
U92436.1 | - | 8 | 1934 | |||
U93051.1 | - | 8 | 900 | |||
U96180.1 | - | 8 | 1704 |