Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317163.1 | NP_001304092.1 | 6 | 963 | ||
NM_001317164.1 | NP_001304093.1 | 6 | 634 | |||
NM_001735.2 | NP_001726.2 | 6 | 637 | |||
XM_011518980.2 | XP_011517282.1 | 7 | 1006 | |||
XM_017015102.1 | XP_016870591.1 | 8 | 1158 | |||
XM_017015103.1 | XP_016870592.1 | 6 | 791 | |||
XM_017015104.1 | XP_016870593.1 | 8 | 1158 | |||
GenBank mRNA | AB209031.1 | - | 6 | 963 | ||
AK310774.1 | - | 6 | 634 | |||
AK310780.1 | - | 6 | 636 | |||
BC022299.1 | - | 6 | 638 | |||
BC113738.1 | - | 6 | 632 | 8322790 | ||
BC113740.1 | - | 6 | 632 | 8322792 | ||
M57729.1 | - | 6 | 619 |