Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291296.1 | NP_001278225.1 | 10 | 1169 | ||
NM_007059.3 | NP_008990.2 | 12 | 1337 | |||
NR_111923.1 | 13 | 1424 | ||||
XM_011526399.2 | XP_011524701.1 | 15 | 2842 | |||
XM_017026226.1 | XP_016881715.1 | 16 | 1804 | |||
XM_017026227.1 | XP_016881716.1 | 15 | 1880 | |||
XM_017026228.1 | XP_016881717.1 | 15 | 1687 | |||
GenBank mRNA | AK023996.1 | - | 12 | 1292 | ||
AK225419.1 | - | 12 | 1281 | |||
AK298672.1 | - | 13 | 1322 | |||
BC009249.2 | - | 12 | 1252 | 3029623 | ||
BG388698.1 | - | 2 | 100 |