Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006059.3 | NP_006050.3 | 2 | 523 | ||
XM_006716921.2 | XP_006716984.1 | 2 | 466 | |||
XM_011518121.1 | XP_011516423.1 | 2 | 466 | |||
GenBank mRNA | AF041835.1 | - | 2 | 522 | ||
AK075536.1 | - | 2 | 466 |