Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000419.4 | NP_000410.2 | 3 | 463 | ||
XM_011524749.1 | XP_011523051.1 | 3 | 367 | |||
XM_011524750.1 | XP_011523052.1 | 3 | 367 | |||
GenBank mRNA | AK297155.1 | - | 3 | 367 | ||
AK315335.1 | - | 3 | 367 | |||
BC117443.1 | - | 3 | 361 | 40125994 | ||
BC126442.1 | - | 3 | 361 | 8992158 | ||
BM148153.1 | - | 3 | 464 | |||
J02764.1 | - | 3 | 336 | |||
M34480.1 | - | 3 | 367 |