Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000295.4 | NP_000286.3 | 5 | 1462 | ||
NM_001002235.2 | NP_001002235.1 | 5 | 1441 | |||
NM_001002236.2 | NP_001002236.1 | 7 | 1755 | |||
NM_001127700.1 | NP_001121172.1 | 5 | 1478 | |||
NM_001127701.1 | NP_001121173.1 | 7 | 1774 | |||
NM_001127702.1 | NP_001121174.1 | 6 | 1582 | |||
NM_001127703.1 | NP_001121175.1 | 7 | 1737 | |||
NM_001127704.1 | NP_001121176.1 | 7 | 1734 | |||
NM_001127705.1 | NP_001121177.1 | 7 | 1752 | |||
NM_001127706.1 | NP_001121178.1 | 6 | 1545 | |||
NM_001127707.1 | NP_001121179.1 | 6 | 1542 | |||
XM_017021370.1 | XP_016876859.1 | 6 | 1642 | |||
GenBank mRNA | AF119873.1 | - | 1413 | |||
AK026174.1 | - | 5 | 1244 | |||
AK315637.1 | - | 6 | 1498 | |||
BC015642.2 | - | 5 | 1228 | 4644658 | ||
BC070163.1 | - | 6 | 1168 | |||
BT019455.1 | - | 4 | 1201 | |||
BX161449.1 | - | 5 | 1368 | |||
BX247968.1 | - | 7 | 1682 | |||
BX248002.1 | - | 4 | 1430 | |||
GU727620.1 | - | 4 | 1202 | |||
K01396.1 | - | 4 | 1220 | |||
KU178257.1 | - | 4 | 1201 | |||
KU178258.1 | - | 2 | 196 | |||
M11465.1 | - | 4 | 1212 | |||
V00496.1 | - | 2 | 155 | |||
X01683.1 | - | 4 | 1239 | |||
X02920.1 | - | 3 | 328 |