Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303110.1 | NP_001290039.1 | 22 | 1500 | ||
NM_001845.5 | NP_001836.3 | 22 | 1500 | |||
XM_011521048.2 | XP_011519350.1 | 22 | 9432 | |||
GenBank mRNA | AB209646.1 | - | 22 | 1485 | ||
BC142626.1 | - | 22 | 1474 | |||
BC151220.1 | - | 22 | 1484 | 40148649 | ||
X05561.1 | - | 22 | 1492 | |||
Y00706.1 | - | 20 | 1235 |