Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPGM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293085.1 | NP_001280014.1 | 3 | 768 | ||
NM_001724.4 | NP_001715.1 | 2 | 643 | |||
NM_199186.2 | NP_954655.1 | 3 | 964 | |||
XM_011516527.1 | XP_011514829.1 | 2 | 824 | |||
GenBank mRNA | AK097778.1 | - | 2 | 567 | ||
AK315439.1 | - | 2 | 611 | |||
BC017050.1 | - | 2 | 614 | 3846430 | ||
BX473275.1 | - | 2 | 627 | |||
X04327.1 | - | 2 | 533 |