Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000505.3 | NP_000496.2 | 6 | 482 | ||
XM_011534462.2 | XP_011532764.1 | 3 | 316 | |||
GenBank mRNA | AB095845.1 | - | 6 | 482 | ||
M11723.1 | - | 6 | 394 | |||
M31315.1 | - | 6 | 424 |