Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006516.2 | NP_006507.2 | 4 | 814 | ||
GenBank mRNA | AB208987.1 | - | 4 | 534 | ||
AK122999.1 | - | 2 | 74 | |||
AK293306.1 | - | 4 | 497 | |||
AK296736.1 | - | 4 | 506 | |||
AK312403.1 | - | 4 | 506 | |||
BC118590.1 | - | 4 | 435 | 40085221 | ||
BC121804.1 | - | 4 | 435 | 40085220 | ||
K03195.1 | - | 4 | 468 |