Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAK3 SIRNA Assays ›
Gene Symbol:
PAK3
Gene Name:
p21 (RAC1) activated kinase 3
Gene Aliases:
ARA, MRX30, MRX47, OPHN3, PAK-3, PAK3beta, bPAK, beta-PAK
Chromosome Location:
Chr.X: 110944241 - 111227361 on Build GRCh38
UniGene ID:
Hs.593599
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534740
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128166.2
NP_001121638.1
7
1070
NM_001128167.2
NP_001121639.1
6
997
NM_001128168.2
NP_001121640.1
11
1319
NM_001128172.2
NP_001121644.1
6
672
NM_001128173.2
NP_001121645.1
10
1256
NM_001324325.1
NP_001311254.1
7
1148
NM_001324326.1
NP_001311255.1
7
993
NM_001324327.1
NP_001311256.1
9
1105
NM_001324328.1
NP_001311257.1
8
1038
NM_001324329.1
NP_001311258.1
10
1253
NM_001324330.1
NP_001311259.1
7
1145
NM_001324331.1
NP_001311260.1
8
1060
NM_001324332.1
NP_001311261.1
9
1211
NM_001324333.1
NP_001311262.1
9
1186
NM_001324334.1
NP_001311263.1
8
1141
NM_002578.4
NP_002569.1
9
1208
NR_136747.1
8
1060
NR_136748.1
6
757
XM_005262132.2
XP_005262189.1
10
1248
XM_005262133.3
XP_005262190.1
9
1203
XM_006724654.2
XP_006724717.1
9
1095
XM_006724655.2
XP_006724718.1
8
1038
XM_011530962.1
XP_011529264.1
10
1246
XM_011530967.2
XP_011529269.1
9
1186
XM_011530968.2
XP_011529270.1
10
1262
XM_011530969.1
XP_011529271.1
8
1107
XM_011530971.2
XP_011529273.1
10
1248
XM_017029557.1
XP_016885046.1
8
1107
XM_017029558.1
XP_016885047.1
8
1050
XM_017029559.1
XP_016885048.1
7
1062
XM_017029560.1
XP_016885049.1
7
993
XM_017029561.1
XP_016885050.1
9
1210
XM_017029562.1
XP_016885051.1
10
1248
XM_017029563.1
XP_016885052.1
11
1311
GenBank mRNA
AB102659.1
-
6
627
AF068864.1
-
5
582
AF155651.1
-
9
973
AK128022.1
-
8
1296
AK290504.1
-
6
997
AK315284.1
-
7
914
AM943850.1
-
6
627
AM943851.1
-
6
645
AM943852.1
-
7
690
BC117353.1
-
6
676
BC117355.1
-
6
676
BC143544.1
-
6
676
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
mitotic cell cycle
stimulatory C-type lectin receptor signaling pathway
axonogenesis
dendrite development
regulation of actin filament polymerization
T cell costimulation
positive regulation of neuron apoptotic process
ephrin receptor signaling pathway
synapse organization
T cell receptor signaling pathway
dendritic spine morphogenesis
positive regulation of dendritic spine morphogenesis
cellular response to organic cyclic compound
positive regulation of DNA biosynthetic process
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Rho GTPase binding
SH3 domain binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0