Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174916.2 | NP_777576.1 | 28 | 3025 | ||
GenBank mRNA | AF061556.1 | - | 10 | 856 | ||
AF525401.1 | - | 28 | 2908 | |||
AK056441.1 | - | 28 | 2975 | |||
AK292516.1 | - | 28 | 2972 | |||
AK302489.1 | - | 28 | 3052 | |||
AY061886.1 | - | 28 | 2959 | |||
BC113505.1 | - | 28 | 3037 | 8322557 | ||
BC113507.1 | - | 28 | 3037 | 8322559 |