Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000157.3 | NP_000148.2 | 4 | 590 | ||
NM_001005741.2 | NP_001005741.1 | 5 | 849 | |||
NM_001005742.2 | NP_001005742.1 | 5 | 830 | |||
NM_001171811.1 | NP_001165282.1 | 3 | 666 | |||
GenBank mRNA | AK291911.1 | - | 5 | 828 | ||
AK300186.1 | - | 510 | ||||
AK300829.1 | - | 3 | 503 | |||
AK300876.1 | - | 5 | 830 | |||
AK302000.1 | - | 3 | 415 | |||
AK311242.1 | - | 5 | 438 | |||
AK312502.1 | - | 5 | 646 | |||
AU131071.1 | - | 5 | 657 | |||
AU140959.1 | - | 4 | 574 | |||
BC003356.1 | - | 4 | 529 | 2899915 | ||
BX382279.2 | - | 6 | 748 | |||
BX648487.1 | - | 5 | 658 | |||
D13286.1 | - | 4 | 529 | |||
K02920.1 | - | 3 | 409 | |||
KJ690771.1 | - | 4 | 473 | |||
KJ690772.1 | - | 4 | 473 | |||
KJ690773.1 | - | 4 | 473 | |||
M16328.1 | - | 4 | 859 | |||
M19285.1 | - | 4 | 546 |