Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL11RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142784.2 | NP_001136256.1 | 13 | 1393 | ||
NR_052010.1 | 13 | 1447 | ||||
GenBank mRNA | AK075530.1 | - | 13 | 1444 | ||
AK094689.1 | - | 13 | 2700 | |||
AK292073.1 | - | 13 | 1427 | |||
BC003110.1 | - | 13 | 1409 | 3502059 | ||
U32324.1 | - | 13 | 1405 | |||
Z38102.1 | - | 12 | 1349 |