Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL11RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142784.2 | NP_001136256.1 | 12, 13 | 1312 | ||
NR_052010.1 | 12, 13 | 1366 | ||||
GenBank mRNA | AK075530.1 | - | 12, 13 | 1363 | ||
AK094689.1 | - | 12, 13 | 2619 | |||
AK292073.1 | - | 12, 13 | 1346 | |||
BC003110.1 | - | 12, 13 | 1328 | 3502059 | ||
U32324.1 | - | 12, 13 | 1324 | |||
Z38102.1 | - | 11, 12 | 1268 |