Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHEJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024782.2 | NP_079058.1 | 5 | 695 | ||
GenBank mRNA | AJ972687.1 | - | 5 | 639 | ||
AK022672.1 | - | 5 | 631 | |||
BC008210.2 | - | 1 | 21 | |||
BC012732.2 | - | 5 | 616 | 4026495 | ||
BC030986.1 | - | 5 | 648 | 4076851 | ||
CR457291.1 | - | 4 | 549 | |||
DB124934.1 | - | 5 | 535 |