Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214711.3 | NP_999876.2 | 3 | 464 | ||
GenBank mRNA | BX484937.1 | - | 3 | 465 | ||
BX640892.1 | - | 3 | 377 | |||
BX640924.1 | - | 3 | 415 | |||
BX640997.1 | - | 3 | 451 | |||
BX647183.1 | - | 3 | 336 | |||
BX647185.1 | - | 4 | 454 | |||
BX648724.1 | - | 3 | 413 | |||
BX648872.1 | - | 3 | 336 | |||
BX648895.1 | - | 3 | 413 |