Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214711.3 | NP_999876.2 | 3 | 379 | ||
GenBank mRNA | BX484937.1 | - | 3 | 380 | ||
BX485867.1 | - | 3 | 376 | |||
BX640892.1 | - | 3 | 292 | |||
BX640924.1 | - | 3 | 330 | |||
BX640997.1 | - | 3 | 366 | |||
BX647183.1 | - | 3 | 251 | |||
BX647185.1 | - | 4 | 369 | |||
BX648724.1 | - | 3 | 328 | |||
BX648872.1 | - | 3 | 251 | |||
BX648895.1 | - | 3 | 328 | |||
BX648896.1 | - | 4 | 263 |