Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004959.4 | NP_004950.2 | 7 | 1496 | ||
XM_005251871.3 | XP_005251928.1 | 7 | 1524 | |||
XM_011518455.2 | XP_011516757.1 | 7 | 1374 | |||
GenBank mRNA | AB307718.1 | - | 6 | 1309 | ||
AK090858.1 | - | 2 | 942 | |||
BC032501.1 | - | 7 | 1496 | 5227690 | ||
HQ692856.1 | - | 6 | 1309 | |||
HQ709184.1 | - | 5 | 1189 | |||
U76388.1 | - | 6 | 1324 |