Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRBN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173482.1 | NP_001166953.1 | 6 | 728 | ||
NM_016302.3 | NP_057386.2 | 6 | 731 | |||
XM_005265202.3 | XP_005265259.1 | 5 | 612 | |||
XM_011533791.2 | XP_011532093.1 | 6 | 718 | |||
XM_011533793.2 | XP_011532095.1 | 7 | 859 | |||
XM_011533794.2 | XP_011532096.1 | 6 | 730 | |||
GenBank mRNA | AF117230.1 | - | 6 | 695 | ||
AK027507.1 | - | 6 | 709 | |||
AK058191.1 | - | 6 | 705 | |||
AK301423.1 | - | 5 | 612 | |||
BC017419.1 | - | 6 | 719 | 4672890 | ||
BC067811.1 | - | 6, 7 | 712 | 5264951 | ||
BX647351.1 | - | 1 | 517 | |||
CR627060.1 | - | 5 | 598 |