Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCSER1 SIRNA Assays ›
Gene Symbol:
CCSER1
Gene Name:
coiled-coil serine rich protein 1
Gene Aliases:
FAM190A
Chromosome Location:
Chr.4: 90127394 - 91602219 on Build GRCh38
UniGene ID:
Hs.654735
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s53491
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCSER1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145065.1
NP_001138537.1
5
1959
NM_207491.2
NP_997374.1
5
1973
XM_011531936.1
XP_011530238.1
6
2194
XM_011531937.2
XP_011530239.1
6
2595
XM_011531938.2
XP_011530240.1
6
1919
XM_011531939.2
XP_011530241.1
6
3419
XM_011531941.2
XP_011530243.1
6
2194
XM_011531942.2
XP_011530244.1
6
2194
XM_011531943.2
XP_011530245.1
6
2194
XM_011531945.1
XP_011530247.1
6
2194
XM_011531946.2
XP_011530248.1
6
2194
XM_011531947.2
XP_011530249.1
6
2194
XM_011531948.1
XP_011530250.1
6
2194
XM_011531949.2
XP_011530251.1
6
2194
XM_011531950.2
XP_011530252.1
6
2194
XM_011531951.2
XP_011530253.1
6
2194
XM_011531955.2
XP_011530257.1
6
2194
XM_011531956.2
XP_011530258.1
6
2194
XM_011531957.2
XP_011530259.1
6
2194
XM_011531958.2
XP_011530260.1
6
2194
XM_017008194.1
XP_016863683.1
6
1917
XM_017008195.1
XP_016863684.1
6
2194
XM_017008196.1
XP_016863685.1
6
2194
XM_017008197.1
XP_016863686.1
5
2098
XM_017008198.1
XP_016863687.1
6
2194
XM_017008199.1
XP_016863688.1
5
2098
GenBank mRNA
AB051467.1
-
5
1907
AK310825.1
-
5
1957
BC051694.1
-
5
1973
5296943
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9bfc7:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline