Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000227.4 | NP_000218.3 | 15 | 1874 | ||
NM_001127717.2 | NP_001121189.2 | 51 | 6749 | |||
NM_001127718.2 | NP_001121190.2 | 14 | 1706 | |||
NM_198129.2 | NP_937762.2 | 52 | 6917 | |||
XM_011525978.2 | XP_011524280.1 | 52 | 6944 | |||
XM_011525979.2 | XP_011524281.1 | 52 | 6935 | |||
XM_011525980.2 | XP_011524282.1 | 52 | 6926 | |||
XM_011525981.2 | XP_011524283.1 | 51 | 6812 | |||
XM_011525982.2 | XP_011524284.1 | 52 | 6944 | |||
XM_017025743.1 | XP_016881232.1 | 35 | 4628 | |||
XM_017025744.1 | XP_016881233.1 | 25 | 3309 | |||
GenBank mRNA | AB107369.1 | - | 52 | 6676 | ||
AB208853.1 | - | 7 | 902 | |||
AY327114.1 | - | 14 | 1681 | |||
AY327115.1 | - | 52 | 6676 | |||
AY327116.1 | - | 51 | 6508 | |||
BC043618.1 | - | 9 | 1254 | |||
L34155.1 | - | 15 | 1816 | |||
X84900.1 | - | 17 | 2095 | |||
X85107.1 | - | 15 | 1867 | |||
X85108.1 | - | 17 | 2095 |