Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000227.4 | NP_000218.3 | 28 | 3666 | ||
NM_001127717.2 | NP_001121189.2 | 64 | 8541 | |||
NM_001127718.2 | NP_001121190.2 | 27 | 3498 | |||
NM_198129.2 | NP_937762.2 | 65 | 8709 | |||
XM_011525978.2 | XP_011524280.1 | 65 | 8736 | |||
XM_011525979.2 | XP_011524281.1 | 65 | 8727 | |||
XM_011525980.2 | XP_011524282.1 | 65 | 8718 | |||
XM_011525981.2 | XP_011524283.1 | 64 | 8604 | |||
XM_011525982.2 | XP_011524284.1 | 63 | 8439 | |||
XM_017025743.1 | XP_016881232.1 | 48 | 6420 | |||
XM_017025744.1 | XP_016881233.1 | 38 | 5101 | |||
GenBank mRNA | AB107369.1 | - | 65 | 8468 | ||
AB208853.1 | - | 20 | 2694 | |||
AY327114.1 | - | 27 | 3473 | |||
AY327115.1 | - | 65 | 8468 | |||
AY327116.1 | - | 64 | 8300 | |||
BC043618.1 | - | 22 | 3046 | |||
L34155.1 | - | 28 | 3608 | |||
X84900.1 | - | 30 | 3887 | |||
X85107.1 | - | 28 | 3659 | |||
X85108.1 | - | 30 | 3887 |