Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127717.2 | NP_001121189.2 | 9 | 1444 | ||
NM_001302996.1 | NP_001289925.1 | 9 | 1444 | |||
NM_198129.2 | NP_937762.2 | 9 | 1444 | |||
NR_130106.1 | 9 | 1444 | ||||
XM_011525978.2 | XP_011524280.1 | 9 | 1444 | |||
XM_011525979.2 | XP_011524281.1 | 9 | 1444 | |||
XM_011525980.2 | XP_011524282.1 | 9 | 1444 | |||
XM_011525981.2 | XP_011524283.1 | 9 | 1444 | |||
XM_011525982.2 | XP_011524284.1 | 9 | 1444 | |||
GenBank mRNA | AB107369.1 | - | 9 | 1203 | ||
AF005258.1 | - | 9 | 1095 | |||
AK096422.1 | - | 9 | 1444 | |||
AY327115.1 | - | 9 | 1203 | |||
AY327116.1 | - | 9 | 1203 | |||
BC033663.1 | - | 6 | 662 | |||
BC093406.1 | - | 9 | 1289 |