Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193465.1 | NP_001180394.1 | 3 | 688 | ||
NM_001193466.1 | NP_001180395.1 | 2 | 902 | |||
NM_015443.3 | NP_056258.1 | 2 | 604 | |||
XM_006721823.1 | XP_006721886.1 | 3 | 688 | |||
XM_006721824.3 | XP_006721887.1 | 3 | 872 | |||
XM_011524628.2 | XP_011522930.1 | 3 | 872 | |||
XM_011524629.2 | XP_011522931.1 | 3 | 872 | |||
XM_011524630.2 | XP_011522932.1 | 3 | 872 | |||
XM_011524631.2 | XP_011522933.1 | 3 | 873 | |||
XM_017024488.1 | XP_016879977.1 | 3 | 688 | |||
XM_017024489.1 | XP_016879978.1 | 2 | 799 | |||
GenBank mRNA | AB033093.1 | - | 1 | 525 | ||
AK291259.1 | - | 2 | 665 | |||
BC098376.1 | - | 2 | 604 | 6181132 | ||
BX538006.1 | - | 2 | 905 |