Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017433.4 | NP_059129.3 | 28 | 3506 | ||
XM_011519498.2 | XP_011517800.1 | 28 | 3350 | |||
XM_011519499.1 | XP_011517801.1 | 28 | 3327 | |||
XM_011519500.2 | XP_011517802.1 | 29 | 3396 | |||
XM_011519502.1 | XP_011517804.1 | 28 | 3350 | |||
XM_011519503.1 | XP_011517805.1 | 28 | 3350 | |||
XM_011519504.1 | XP_011517806.1 | 28 | 3350 | |||
XM_011519505.1 | XP_011517807.1 | 28 | 3350 | |||
XM_011519506.2 | XP_011517808.1 | 28 | 3350 | |||
XM_011519508.2 | XP_011517810.1 | 28 | 3350 | |||
XM_011519510.1 | XP_011517812.1 | 28 | 3350 | |||
XM_011519512.1 | XP_011517814.1 | 11 | 1354 | |||
XM_011519513.2 | XP_011517815.1 | 9 | 1708 | |||
GenBank mRNA | AF229172.1 | - | 28 | 3321 | ||
AY101367.1 | - | 28 | 3324 | |||
BC125272.1 | - | 26 | 3079 |