Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IRAK4 SIRNA Assays ›
Gene Symbol:
IRAK4
Gene Name:
interleukin 1 receptor associated kinase 4
Gene Aliases:
IPD1, IRAK-4, NY-REN-64, REN64
Chromosome Location:
Chr.12: 43758913 - 43789543 on Build GRCh38
UniGene ID:
Hs.138499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s535020
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IRAK4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114182.2
NP_001107654.1
6
725
NM_001145256.1
NP_001138728.1
5
579
NM_001145257.1
NP_001138729.1
4
531
NM_001145258.1
NP_001138730.1
3
361
NM_016123.3
NP_057207.2
5
677
XM_005268943.3
XP_005269000.1
5
782
XM_005268944.4
XP_005269001.1
5
746
XM_005268945.4
XP_005269002.1
5
990
XM_005268947.4
XP_005269004.1
6
736
XM_005268948.2
XP_005269005.1
7
776
XM_005268949.2
XP_005269006.1
4
636
XM_006719438.3
XP_006719501.1
6
1038
XM_006719439.2
XP_006719502.1
5
1143
XM_011538431.2
XP_011536733.1
6
794
XM_011538432.1
XP_011536734.1
6
830
XM_011538433.2
XP_011536735.1
5
1409
XM_017019390.1
XP_016874879.1
7
911
XM_017019391.1
XP_016874880.1
6
876
GenBank mRNA
AF155118.1
-
5
644
AF445802.1
-
4
595
AK000528.1
-
5
624
AK027301.2
-
5
1172
AK299944.1
-
4
528
AK304583.1
-
3
364
AK314288.1
-
5
666
AY283670.1
-
4
595
AY283671.1
-
4
595
AY340962.1
-
5
660
AY340963.1
-
6
708
AY340964.1
-
4
514
AY340965.1
-
5
562
AY340966.1
-
6
727
AY340967.1
-
7
775
AY730682.1
-
4
595
BC013316.1
-
5
665
4287014
KU178631.1
-
4
595
KU178632.1
-
3
449
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
toll-like receptor signaling pathway
neutrophil mediated immunity
MyD88-dependent toll-like receptor signaling pathway
JNK cascade
cytokine-mediated signaling pathway
toll-like receptor 9 signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
positive regulation of smooth muscle cell proliferation
neutrophil migration
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
interleukin-1 receptor binding
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-558c998fd6-99mnp:80/10.239.163.72:80.
git-commit: a0437b286f8214373ac979cfd76c0df1857d95b8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.14.2-2024.08.97-1.0