Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F13A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000129.3 | NP_000120.2 | 5 | 741 | ||
GenBank mRNA | AB208852.1 | - | 5 | 693 | ||
AK304335.1 | - | 4 | 523 | |||
AK312735.1 | - | 5 | 712 | |||
BC027963.1 | - | 5 | 699 | 5222962 | ||
M14354.1 | - | 5 | 702 | |||
M14539.1 | - | 5 | 705 |