Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F13A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000129.3 | NP_000120.2 | 15 | 2297 | ||
GenBank mRNA | AB208852.1 | - | 15 | 2249 | ||
AK130666.1 | - | 8 | 1143 | |||
AK304335.1 | - | 14 | 1950 | |||
AK312735.1 | - | 15 | 2268 | |||
BC027963.1 | - | 15 | 2255 | 5222962 | ||
M14354.1 | - | 15 | 2258 | |||
M14539.1 | - | 15 | 2261 |