Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNMT3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207055.1 | NP_001193984.1 | 6, 7 | 1013 | ||
NM_001207056.1 | NP_001193985.1 | 5, 6 | 911 | |||
NM_006892.3 | NP_008823.1 | 7, 8 | 1139 | |||
NM_175848.1 | NP_787044.1 | 7, 8 | 1139 | |||
NM_175849.1 | NP_787045.1 | 7, 8 | 1139 | |||
NM_175850.2 | NP_787046.1 | 7, 8 | 1118 | |||
XM_011528653.2 | XP_011526955.1 | 7, 8 | 991 | |||
XM_011528654.2 | XP_011526956.1 | 6, 7 | 866 | |||
GenBank mRNA | AB208880.1 | - | 7, 8 | 1117 | ||
AF156487.1 | - | 7, 8 | 932 | |||
AF156488.1 | - | 7, 8 | 932 | |||
AF176228.1 | - | 7, 8 | 1118 | |||
AF331857.1 | - | 7, 8 | 1139 | |||
AK001191.1 | - | 11 | ||||
AK295176.1 | - | 6, 7 | 780 | |||
AK299776.1 | - | 6, 7 | 752 | |||
AK299821.1 | - | 5, 6 | 654 | |||
AK299915.1 | - | 6, 7 | 790 | |||
AK300857.1 | - | 6, 7 | 802 | |||
DQ321787.1 | - | 7, 8 | 1139 |