Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS7BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029875.2 | NP_001025046.1 | 5 | 1334 | ||
NM_001271890.1 | NP_001258819.1 | 3 | 448 | |||
NM_001271891.1 | NP_001258820.1 | 3 | 446 | |||
NR_073511.1 | 3 | 448 | ||||
XM_005248502.3 | XP_005248559.1 | 5 | 2714 | |||
GenBank mRNA | AK294808.1 | - | 4 | 439 | ||
AK296443.1 | - | 5 | 982 | |||
AK296494.1 | - | 3 | 416 | |||
BC140707.1 | - | 5 | 680 | 9021575 | ||
BX640900.1 | - | 5 | 572 | |||
DA211241.1 | - | 3 | 449 | |||
DA417118.1 | - | 3 | 416 |