Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLEC7A SIRNA Assays ›
Gene Symbol:
CLEC7A
Gene Name:
C-type lectin domain family 7 member A
Gene Aliases:
BGR, CANDF4, CD369, CLECSF12, DECTIN1, SCARE2
Chromosome Location:
Chr.12: 10116777 - 10130269 on Build GRCh38
UniGene ID:
Hs.143929
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s535050
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLEC7A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022570.4
NP_072092.2
1
225
NM_197947.2
NP_922938.1
1
225
NM_197948.2
NP_922939.1
1
225
NM_197949.2
NP_922940.1
1
225
NM_197950.2
NP_922941.1
1
225
NM_197954.2
NP_922945.1
1
225
NR_125336.1
1
225
XM_006719135.3
XP_006719198.1
1
220
XM_017019822.1
XP_016875311.1
1
219
XM_017019823.1
XP_016875312.1
1
122
GenBank mRNA
AA361063.1
-
1
60
AF313468.1
-
2
89
AF313469.1
-
1
88
AF400595.1
-
1
38
AF400596.1
-
1
38
AF400597.1
-
1
38
AF400598.1
-
1
38
AF400599.1
-
1
38
AF400600.1
-
1
38
AF400601.1
-
1
38
AF400602.1
-
1
38
AJ312372.1
-
1
193
AJ312373.1
-
1
193
AK294111.1
-
1
217
AK295108.1
-
1
169
AK297028.1
-
1
109
AK298679.1
-
1
141
AK298724.1
-
1
141
AK313247.1
-
1
156
AY009090.2
-
1
109
AY026769.2
-
1
38
AY026770.2
-
1
38
AY026771.1
-
1
38
AY359002.1
-
1
197
BC013385.1
-
1
94
BC071746.1
-
1
139
4690475
BC093829.1
-
1
114
BC093831.1
-
1
114
CR749536.1
-
1
168
DA193873.1
-
1
225
KU178745.1
-
1
38
KU178746.1
-
1
38
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
pattern recognition receptor signaling pathway
stimulatory C-type lectin receptor signaling pathway
leukocyte activation involved in immune response
phagocytosis, recognition
inflammatory response
cell recognition
carbohydrate mediated signaling
T cell activation
defense response to protozoan
innate immune response
cellular response to molecule of fungal origin
protein binding
signaling pattern recognition receptor activity
carbohydrate binding
MHC protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-ndlhf:80/100.66.76.213:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline