Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000286.2 | NP_000277.1 | 3 | 1644 | ||
GenBank mRNA | AB004546.1 | - | 3 | 1213 | ||
AK312635.1 | - | 3 | 1557 | |||
BC031085.1 | - | 3 | 1548 | 5296541 | ||
U91521.1 | - | 3 | 1575 |