Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145102.1 | NP_001138574.1 | 6 | 766 | ||
NM_001145103.1 | NP_001138575.1 | 6 | 577 | |||
NM_001145104.1 | NP_001138576.1 | 4 | 210 | |||
NM_005902.3 | NP_005893.1 | 6 | 1001 | |||
XM_011521559.2 | XP_011519861.1 | 5 | 1028 | |||
XM_011521560.1 | XP_011519862.1 | 6 | 940 | |||
GenBank mRNA | AB209616.1 | - | 6 | 586 | ||
AK290881.1 | - | 7 | 926 | |||
AK298139.1 | - | 6 | 577 | |||
AK300614.1 | - | 4 | 207 | |||
AK301348.1 | - | 4 | 202 | |||
AK316017.1 | - | 6 | 626 | |||
BC050743.1 | - | 6 | 919 | 6094328 | ||
DA755047.1 | - | 1 | 51 | |||
DA977882.1 | - | 4 | 210 | |||
EU016553.1 | - | 4 | 215 | |||
U68019.1 | - | 6 | 769 | |||
U76622.1 | - | 6 | 740 |