Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145102.1 | NP_001138574.1 | 7, 8 | 1081 | ||
NM_001145103.1 | NP_001138575.1 | 7, 8 | 892 | |||
NM_001145104.1 | NP_001138576.1 | 5, 6 | 525 | |||
NM_005902.3 | NP_005893.1 | 7, 8 | 1316 | |||
XM_011521559.2 | XP_011519861.1 | 6, 7 | 1343 | |||
XM_011521560.1 | XP_011519862.1 | 7, 8 | 1255 | |||
GenBank mRNA | AB209616.1 | - | 7, 8 | 901 | ||
AK026690.1 | - | 1 | 714 | |||
AK290881.1 | - | 8, 9 | 1241 | |||
AK298139.1 | - | 7, 8 | 892 | |||
AK300614.1 | - | 5, 6 | 522 | |||
AK316017.1 | - | 7, 8 | 941 | |||
BC050743.1 | - | 7, 8 | 1234 | 6094328 | ||
DA755047.1 | - | 2, 3 | 366 | |||
DA977882.1 | - | 5, 6 | 525 | |||
U68019.1 | - | 7, 8 | 1084 | |||
U76622.1 | - | 7, 8 | 1055 |