Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015338.5 | NP_056153.2 | 13 | 3068 | ||
XM_006723727.3 | XP_006723790.1 | 12 | 3347 | |||
XM_006723728.3 | XP_006723791.1 | 13 | 2677 | |||
XM_006723730.3 | XP_006723793.1 | 13 | 2873 | |||
XM_006723732.2 | XP_006723795.1 | 12 | 2696 | |||
XM_006723733.1 | XP_006723796.1 | 6 | 2702 | |||
XM_011528648.2 | XP_011526950.1 | 13 | 2992 | |||
XM_011528652.2 | XP_011526954.1 | 14 | 4010 | |||
XM_017027704.1 | XP_016883193.1 | 13 | 4133 | |||
XM_017027705.1 | XP_016883194.1 | 13 | 3146 | |||
XM_017027706.1 | XP_016883195.1 | 12 | 3197 | |||
GenBank mRNA | AB023195.2 | - | 7 | 2118 | ||
AB084280.1 | - | 12 | 2621 | |||
AJ438952.2 | - | 13 | 2894 | |||
AK122923.1 | - | 13 | 2642 | |||
AL117518.1 | - | 6 | 2038 | |||
BC137278.1 | - | 14 | 2831 | 9021275 | ||
BC137280.1 | - | 14 | 2831 | 9021277 |