Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006642.3 | NP_006633.1 | 12 | 1549 | ||
XM_005273013.4 | XP_005273070.1 | 11 | 1381 | |||
XM_005273018.1 | XP_005273075.1 | 11 | 1388 | |||
XM_005273021.4 | XP_005273078.1 | 12 | 1321 | |||
XM_005273022.3 | XP_005273079.1 | 9 | 1170 | |||
XM_006711727.3 | XP_006711790.1 | 12 | 1435 | |||
XM_006711728.3 | XP_006711791.1 | 11 | 1306 | |||
XM_011544021.2 | XP_011542323.1 | 13 | 1531 | |||
XM_011544022.2 | XP_011542324.1 | 13 | 1606 | |||
XM_011544023.2 | XP_011542325.1 | 13 | 1531 | |||
XM_011544024.2 | XP_011542326.1 | 13 | 1531 | |||
XM_011544026.2 | XP_011542328.1 | 13 | 1531 | |||
XM_011544028.2 | XP_011542330.1 | 11 | 1306 | |||
XM_011544030.2 | XP_011542332.1 | 5 | 2082 | |||
XM_017000103.1 | XP_016855592.1 | 13 | 1531 | |||
XM_017000104.1 | XP_016855593.1 | 11 | 1381 | |||
XM_017000105.1 | XP_016855594.1 | 12 | 1536 | |||
XM_017000106.1 | XP_016855595.1 | 12 | 1517 | |||
XM_017000107.1 | XP_016855596.1 | 13 | 1433 | |||
XM_017000108.1 | XP_016855597.1 | 15 | 1980 | |||
GenBank mRNA | AF039690.1 | - | 6 | 720 | ||
AF250731.1 | - | 12 | 1380 | |||
AK297105.1 | - | 8 | 968 | |||
AK299212.1 | - | 12 | 1498 | |||
AK299249.1 | - | 9 | 1141 |