Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308093.1 | NP_001295022.1 | 6 | 1836 | ||
NM_001308094.1 | NP_001295023.1 | 7 | 1176 | |||
NM_002052.4 | NP_002043.2 | 7 | 1987 | |||
XM_005272385.4 | XP_005272442.1 | 8 | 2059 | |||
XM_005272386.1 | XP_005272443.1 | 8 | 2247 | |||
XM_006716248.1 | XP_006716311.1 | 8 | 2168 | |||
XM_011543817.2 | XP_011542119.1 | 8 | 2238 | |||
XM_011543818.2 | XP_011542120.1 | 7 | 2376 | |||
XM_017013312.1 | XP_016868801.1 | 7 | 2692 | |||
GenBank mRNA | AK097060.1 | - | 7 | 1136 | ||
D78260.1 | - | 6 | 1668 | |||
L34357.1 | - | 6 | 1616 |