Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308093.1 | NP_001295022.1 | 1, 2 | 1074 | ||
XM_005272385.4 | XP_005272442.1 | 3, 4 | 1297 | |||
XM_005272386.1 | XP_005272443.1 | 3, 4 | 1485 | |||
XM_006716248.1 | XP_006716311.1 | 3, 4 | 1406 | |||
XM_011543817.2 | XP_011542119.1 | 3, 4 | 1476 | |||
XM_011543818.2 | XP_011542120.1 | 2, 3 | 1614 | |||
XM_017013312.1 | XP_016868801.1 | 2, 3 | 1930 | |||
GenBank mRNA | BC143479.1 | - | 1, 2 | 697 | 9051987 |