Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT1A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019075.2 | NP_061948.1 | 1 | 303 | ||
GenBank mRNA | AY435137.1 | - | 1 | 257 | ||
BC020971.1 | - | 1 | 326 | 3850654 | ||
BC053576.1 | - | 1 | 278 | 4452440 | ||
BC069210.1 | - | 1 | 287 | 4660540 | ||
U89508.1 | - | 1 | 257 |